Learn More
Biomatik Corporation Phospho-IKK-γ (Ser85) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51361
Encompass_Preferred
Phospho-IKK-γ (Ser85) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IKK-γ (Ser85) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9Y6K9. Gene ID: 8517. Target Synonyms: NEMO; NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase subunit gamma; I-kappa-B kinase gamma; IKK-gamma; IKKG; IkB kinase-associated protein 1; IKKAP1; FIP-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1522