Learn More
Biomatik Corporation Phospho-MRE11 (Ser264) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51377
Encompass_Preferred
Phospho-MRE11 (Ser264) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MRE11 (Ser264) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49959. Gene ID: 4361. Target Synonyms: Double-strand break repair protein MRE11A; HNGS1; MRE11; MRE11 homolog 1; MRE11A; MmMRE11A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1536