Learn More
Biomatik Corporation Phospho-Vitamin D Receptor (Ser208) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51387
Encompass_Preferred
Phospho-Vitamin D Receptor (Ser208) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Vitamin D Receptor (Ser208) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11473. Gene ID: 7421. Target Synonyms: 1,25-dihydroxyvitamin D3 receptor; NR1I1; vitamin D receptor; vitamin D3 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1545