Learn More
Biomatik Corporation Phospho-ATF2 (Thr691) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51397
Encompass_Preferred
Phospho-ATF2 (Thr691) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ATF2 (Thr691) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15336. Gene ID: 1386. Target Synonyms: Activating 2; ATF2; cAMP response element binding protein CRE- BP1; CREB2; CREBP1; Cyclic-AMP-dependent ATF-2; HB16; MXBP protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1553