Learn More
Biomatik Corporation Phospho-HER2 (Tyr877) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51471
Encompass_Preferred
Phospho-HER2 (Tyr877) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HER2 (Tyr877) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04626. Gene ID: 2064. Target Synonyms: Receptor tyrosine-protein kinase erbB-2; Metastatic lymph node gene 19 protein; MLN 19; Proto-oncogene Neu; Proto-oncogene c-ErbB-2; Tyrosine kinase-type cell surface receptor HER2; p185erbB2; CD340; ERBB2; HER2; MLN19; NEU; NGL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1611