Learn More
Biomatik Corporation Phospho-HSP27 (Ser78) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51476
Encompass_Preferred
Phospho-HSP27 (Ser78) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HSP27 (Ser78) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04792. Gene ID: 3315. Target Synonyms: 28 kDa heat shock protein; Estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; Heat shock 27 kDa protein; HSP 27; HSP25; HSPB1; P25; SRP27; Stress-responsive protein 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1615