Learn More
Biomatik Corporation Phospho-JAK1 (Tyr1022) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51486
Encompass_Preferred
Phospho-JAK1 (Tyr1022) Colorimetric Cell-Based ELISA Kit for detecting Phospho-JAK1 (Tyr1022) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23458. Gene ID: 3716. Target Synonyms: JAK-1; JAK1; JAK1A; Janus kinase 1; kinase Jak1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1623