Learn More
Biomatik Corporation Phospho-FANCD2 (Ser222) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51066
Encompass_Preferred
Phospho-FANCD2 (Ser222) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FANCD2 (Ser222) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9BXW9. Gene ID: 2177. Target Synonyms: FACD2; FANCD2; Fanconi anemia complementation group D2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9010