Learn More
Biomatik Corporation Phospho-p95-NBS1 (Ser343) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51514
Encompass_Preferred
Phospho-p95-NBS1 (Ser343) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p95-NBS1 (Ser343) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60934. Gene ID: 4683. Target Synonyms: Cell cycle regulatory protein P95; NBN; NBS; NIBRIN; NIJMEGEN BREAKAGE syndrome protein 1; p95-NBS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1649