Learn More
Biomatik Corporation Phospho-PDK1 (Ser241) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51545
Encompass_Preferred
Phospho-PDK1 (Ser241) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PDK1 (Ser241) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15530. Gene ID: 5170. Target Synonyms: 3-phosphoinositide dependent protein kinase-1; hPDK1; kinase PDK1; mPDK1; PDPK1; PkB kinase; Protein kinase B kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1677