Learn More
Biomatik Corporation Phospho-HSP90B (Ser254) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50975
Encompass_Preferred
Phospho-HSP90B (Ser254) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HSP90B (Ser254) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08238. Gene ID: 3326. Target Synonyms: HS90B; HSP 84; HSP90-beta; HSP90AB1; HSPC2; HSPCB; Heat shock protein HSP 90-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1191