Learn More
Biomatik Corporation Phospho-MEF2A (Ser408) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50981
Encompass_Preferred
Phospho-MEF2A (Ser408) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MEF2A (Ser408) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02078. Gene ID: 4205. Target Synonyms: MEF2; Myocyte-specific enhancer factor 2A; Serum response factor-like protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1196