Learn More
Biomatik Corporation Phospho-p70 S6 Kinase (Thr229) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51132
Encompass_Preferred
Phospho-p70 S6 Kinase (Thr229) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p70 S6 Kinase (Thr229) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23443. Gene ID: 6198. Target Synonyms: Ribosomal protein S6 kinase I; S6K; S6K1; 70 kDa ribosomal protein S6 kinase 1; p70 S6 kinase alpha; p70(S6K)-alpha; p70-S6K; P70S6K; p70-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9020