Learn More
Biomatik Corporation Phospho-CCR5 (Ser349) Colorimetric Cell-Based ELISA Kit
Supplier: Biomatik Corporation EKA51013
Phospho-CCR5 (Ser349) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CCR5 (Ser349) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P51681. Gene ID: 1234; 727797. Target Synonyms: C-C CKR-5; C-C chemokine receptor type 5; CC-CKR-5; CCR-5; CD195 antigen; CHEMR13; CKR5; CMKBR5; HIV-1 fusion co-receptor; MIP-1 alpha receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.