Learn More
Biomatik Corporation Phospho-TIE2 (Tyr1108) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51164
Encompass_Preferred
Phospho-TIE2 (Tyr1108) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TIE2 (Tyr1108) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02763. Gene ID: 7010. Target Synonyms: Tyrosine-protein kinase receptor TIE-2; hTIE2; Tyrosine-protein kinase receptor TEK; p140 TEK; Tunica interna endothelial cell kinase; CD202b antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9024