Learn More
Biomatik Corporation Phospho-BCAR1 (Tyr410) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51025
Encompass_Preferred
Phospho-BCAR1 (Tyr410) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BCAR1 (Tyr410) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P56945. Gene ID: 9564. Target Synonyms: BCAR1; Breast cancer anti-estrogen resistance 1; CRK-ASSOCIATED SUBSTRATE; CRKAS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1231