Learn More
Biomatik Corporation Phospho-PLCβ3 (Ser1105) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51026
Encompass_Preferred
Phospho-PLCβ3 (Ser1105) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PLCβ3 (Ser1105) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q01970. Gene ID: 5331. Target Synonyms: 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 3; Phosphoinositide phospholipase C; Phospholipase C-beta-3; PLC-beta-3; PLCB3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1232