Learn More
Biomatik Corporation Phospho-Chk2 (Thr387) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51056
Encompass_Preferred
Phospho-Chk2 (Thr387) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Chk2 (Thr387) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O96017. Gene ID: 11200. Target Synonyms: Serine/threonine-protein kinase Chk2; Cds1; CHEK2; CHK2; RAD53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1257