Learn More
Biomatik Corporation Phospho-Opioid Receptor-δ (Ser363) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51061
Encompass_Preferred
Phospho-Opioid Receptor-δ (Ser363) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Opioid Receptor-δ (Ser363) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41143. Gene ID: 4985. Target Synonyms: Delta-opioid receptor; Delta-type opioid receptor; K56; MSL-2; OPRD; OPRD1; Opioid receptor delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1262