Learn More
Biomatik Corporation Phospho-Ku70/XRCC6 (Ser5) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51084
Encompass_Preferred
Phospho-Ku70/XRCC6 (Ser5) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Ku70/XRCC6 (Ser5) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12956. Gene ID: 2547. Target Synonyms: X-ray repair cross-complementing protein 6; 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; X-ray repair complementing defective repair in Chinese hamster cells 6; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; Lupus Ku autoantigen protein p70; Ku70; 70 kDa subunit of Ku antigen; Thyroid-lupus autoantigen; TLAA; CTC box-binding factor 75 kDa subunit; CTCBF; CTC75; DNA repair protein XRCC6; XRCC6; G22P1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1283