Learn More
Biomatik Corporation Phospho-Akt1 S1 (Thr246) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51086
Encompass_Preferred
Phospho-Akt1 S1 (Thr246) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Akt1 S1 (Thr246) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96B36. Gene ID: 84335. Target Synonyms: Akt1 substrate 1 (proline-rich) AKTS1; Lobe; MGC2865; PRAS40; proline-rich Akt substrate 40 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1285