Learn More
Biomatik Corporation Phospho-CBL (Tyr700) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51094
Encompass_Preferred
Phospho-CBL (Tyr700) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CBL (Tyr700) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22681. Gene ID: 867. Target Synonyms: Cas-Br-M (murine) ecotropic retroviral transforming; CBL E3 ubiquitin protein ligase; CBL2; Proto-oncogene c-CBL; Signal transduction protein CBL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1292