Learn More
Biomatik Corporation Phospho-JNK1/2/3 (Thr183+Tyr185) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51115
Encompass_Preferred
Phospho-JNK1/2/3 (Thr183+Tyr185) Colorimetric Cell-Based ELISA Kit for detecting Phospho-JNK1/2/3 (Thr183+Tyr185) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P45983; P45984; P53779. Gene ID: 5599; 5601; 5602. Target Synonyms: JNK-46; JNK1-alpha-2; MAPK8; MK08; Mitogen-activated protein kinase 8; PRKM8; SAPK gamma; Stress-activated protein kinase JNK1; c-Jun N-terminal kinase 1; kinase JNK1; p54 gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1310