Learn More
Biomatik Corporation Phospho-SOX-9 (Ser181) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51157
Encompass_Preferred
Phospho-SOX-9 (Ser181) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SOX-9 (Ser181) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48436. Gene ID: 6662. Target Synonyms: SRY sex determining region Y-box 9 protein; SRY-box containing gene 9; Sox-9; transcription factor SOX-9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1344