Learn More
Biomatik Corporation Phospho-Tyrosine Hydroxylase (Ser8) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51169
Encompass_Preferred
Phospho-Tyrosine Hydroxylase (Ser8) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Tyrosine Hydroxylase (Ser8) in researchers plate their cell line of choice. Species Reactivity: Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04177. Gene ID: 25085. Target Synonyms: TH isoform 3; TH isoform a; TH-4; TY3H; TYH; Tyrosine 3-hydroxylase; Tyrosine 3-monooxygenase; tyrosine hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1354