Learn More
Biomatik Corporation Phospho-Akt1 (Thr72) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51178
Encompass_Preferred
Phospho-Akt1 (Thr72) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Akt1 (Thr72) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P31749. Gene ID: 207. Target Synonyms: AKT; AKT1 kinase; C-AKT; kinase Akt1; PKB; PKB-alpha; Protein kinase B; RAC; RAC-alpha serine/threonine kinase; RAC-PK-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1360