Learn More
Biomatik Corporation Phospho-MEK1 (Ser298) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51197
Encompass_Preferred
Phospho-MEK1 (Ser298) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MEK1 (Ser298) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02750. Gene ID: 5604. Target Synonyms: Dual specificity mitogen-activated protein kinase kinase 1; ERK activator kinase 1; MAP kinase kinase 1; MAP2K1; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK1; MP2K1; PRKMK1; kinase MEK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1376