Learn More
Biomatik Corporation Phospho-Mst1/2 (Thr183) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51200
Encompass_Preferred
Phospho-Mst1/2 (Thr183) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Mst1/2 (Thr183) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13188; Q13043. Gene ID: 6789. Target Synonyms: STE20-like kinase MST1; Yeast Sps1/Ste20-related kinase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1379