Learn More
Biomatik Corporation Phospho-PEA-15 (Ser104) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51209
Encompass_Preferred
Phospho-PEA-15 (Ser104) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PEA-15 (Ser104) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15121. Gene ID: 8682. Target Synonyms: Astrocytic phosphoprotein PEA-15; PE15; PEA15; PED; Phosphoprotein enriched in diabetes. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1387