Learn More
Biomatik Corporation Phospho-PKD1/2/3 (Ser738+Ser742) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51215
Encompass_Preferred
Phospho-PKD1/2/3 (Ser738+Ser742) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKD1/2/3 (Ser738+Ser742) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15139; Q9BZL6; O94806. Gene ID: 5587; 25865; 23683. Target Synonyms: KPCD1; PKC-mu; PKCM; PKD; PRKCM; PRKD1; Protein kinase C mu type; Protein kinase D; kinase PKD1; nPKC-mu. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1391