Learn More
Biomatik Corporation Phospho-PFKFB2 (Ser483) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51223
Encompass_Preferred
Phospho-PFKFB2 (Ser483) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PFKFB2 (Ser483) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60825. Gene ID: 5208. Target Synonyms: 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2; 6PF-2-K/Fru-2,6-P2ase 2; PFK/FBPase 2; 6PF-2-K/Fru-2,6-P2ase heart-type isozyme; PFKFB2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1398