Learn More
Biomatik Corporation Phospho-A-RAF (Tyr302) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51227
Encompass_Preferred
Phospho-A-RAF (Tyr302) Colorimetric Cell-Based ELISA Kit for detecting Phospho-A-RAF (Tyr302) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10398. Gene ID: 369. Target Synonyms: A-Raf proto-oncogene serine/threonine-protein kinase; A-raf-1; Proto-oncogene Pks; ARAF; ARAF1; PKS; PKS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1401