Learn More
Biomatik Corporation Phospho-BCL-XL (Thr47) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51229
Encompass_Preferred
Phospho-BCL-XL (Thr47) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BCL-XL (Thr47) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q07817. Gene ID: 598. Target Synonyms: Apoptosis regulator Bcl-X; Bcl-2-like 1 protein; BCL2L; BCL2L1; BCLX; BCLXL; BLC2L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1403