Learn More
Biomatik Corporation Phospho-TIP60 (Ser86) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51249
Encompass_Preferred
Phospho-TIP60 (Ser86) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TIP60 (Ser86) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92993. Gene ID: 10524. Target Synonyms: 60 kDa Tat interactive protein; cPLA(2)-interacting protein; histone acetyltransferase HTATIP; HIV-1 Tat interactive protein; HTATIP; Tat-interactive 60 kDa protein alpha isoform; Tat-interactive protein-60; TI60. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1421