Learn More
Biomatik Corporation Phospho-CDC2 (Tyr15) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51412
Encompass_Preferred
Phospho-CDC2 (Tyr15) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC2 (Tyr15) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06493. Gene ID: 983. Target Synonyms: CDC28; CDC2A; CDK1; cell division control protein 2 homolog; cyclin-dependent kinase 1; kinase Cdc2; MPF; p34 protein kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9053