Learn More
Biomatik Corporation Phospho-Calcium Sensing Receptor (Thr888) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51259
Encompass_Preferred
Phospho-Calcium Sensing Receptor (Thr888) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Calcium Sensing Receptor (Thr888) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41180. Gene ID: 846. Target Synonyms: Extracellular calcium-sensing receptor; CaSR; Parathyroid Cell calcium-sensing receptor; PCaR1; CASR; GPRC2A; PCAR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1431