Learn More
Biomatik Corporation Phospho-CaMK1-α (Thr177) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51261
Encompass_Preferred
Phospho-CaMK1-α (Thr177) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CaMK1-α (Thr177) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14012. Gene ID: 8536. Target Synonyms: CAM kinase I; CAMK1; Calcium/calmodulin-dependent protein kinase type I; KCC1; KCC1A; kinase CaMK1-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1433