Learn More
Biomatik Corporation Phospho-CD5 (Tyr453) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51269
Encompass_Preferred
Phospho-CD5 (Tyr453) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CD5 (Tyr453) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06127. Gene ID: 921. Target Synonyms: LEU1; Lymphocyte antigen CD5; Lymphocyte glycoprotein Ly-1; Lymphocyte glycoprotein T1/Leu-1; Lyt-1; T-cell surface glycoprotein CD5 precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1440