Learn More
Biomatik Corporation Phospho-CD71/TfR (Ser24) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51271
Encompass_Preferred
Phospho-CD71/TfR (Ser24) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CD71/TfR (Ser24) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P02786. Gene ID: 4155. Target Synonyms: CD71 antigen; T9; TFRC; TR; TfR1; Transferrin receptor protein 1; Trfr; p90. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1442