Learn More
Biomatik Corporation Phospho-CDC25B (Ser353) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51273
Encompass_Preferred
Phospho-CDC25B (Ser353) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC25B (Ser353) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P30305. Gene ID: 994. Target Synonyms: CDC25HU2; CDC25M2; Dual specificity phosphatase Cdc25B; M-phase inducer phosphatase 2; MPIP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1444