Learn More
Biomatik Corporation Phospho-Coagulation Factor III (Ser290) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51278
Encompass_Preferred
Phospho-Coagulation Factor III (Ser290) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Coagulation Factor III (Ser290) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P13726. Gene ID: 2152. Target Synonyms: CD142 antigen; F3; Tissue factor; coagulation factor III; thromboplastin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1448