Learn More
Biomatik Corporation Phospho-Catenin-δ1 (Tyr228) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51284
Encompass_Preferred
Phospho-Catenin-δ1 (Tyr228) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Catenin-δ1 (Tyr228) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60716. Gene ID: 1500. Target Synonyms: Catenin delta-1; Cadherin-associated Src substrate; CAS; p120 catenin; p120(ctn) p120(cas) CTNND1; KIAA0384. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1452