Learn More
Biomatik Corporation Phospho-EPHA2/3/4 (Tyr588/596) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51293
Encompass_Preferred
Phospho-EPHA2/3/4 (Tyr588/596) Colorimetric Cell-Based ELISA Kit for detecting Phospho-EPHA2/3/4 (Tyr588/596) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29317; P29320; P54764. Gene ID: 1969; 2042; 2043. Target Synonyms: ECK; Ephrin type-A receptor 2 precursor; Epithelial cell kinase; MPK-5; SEK-2; SEK2; Tyrosine-protein kinase receptor ECK; kinase EphA2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1461