Learn More
Biomatik Corporation Phospho-GATA4 (Ser262) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51300
Encompass_Preferred
Phospho-GATA4 (Ser262) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GATA4 (Ser262) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P43694. Gene ID: 2626. Target Synonyms: DNA binding protein GATA-GT2; GAT4; GATA binding factor-4; GATA-4; Transcription factor GATA-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1467