Learn More
Biomatik Corporation Phospho-DARPP-32 (Thr34) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51427
Encompass_Preferred
Phospho-DARPP-32 (Thr34) Colorimetric Cell-Based ELISA Kit for detecting Phospho-DARPP-32 (Thr34) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UD71. Gene ID: 84152. Target Synonyms: Dopamine- and cAMP-regulated neuronal phosphoprotein; IPPD; Neuronal phosphoprotein DARPP-32; PPP1R1B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9058