Learn More
Biomatik Corporation Phospho-IL-13Rα1 (Tyr405) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51339
Encompass_Preferred
Phospho-IL-13Rα1 (Tyr405) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IL-13Rα1 (Tyr405) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P78552. Gene ID: 3597. Target Synonyms: CD213a1 antigen; I13R1; IL- 13RA-1; IL-13R alpha 1; IL-13R-alpha-1; IL13RA; IL13RA1; Interleukin-13 receptor alpha-1 chain precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1503