Learn More
Biomatik Corporation Phospho-ADD1/2 (Ser726) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50966
Encompass_Preferred
Phospho-ADD1/2 (Ser726) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ADD1/2 (Ser726) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35611; P35612. Gene ID: 118; 119. Target Synonyms: Alpha-adducin; Erythrocyte adducin subunit alpha; ADD1; ADDA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1183