Learn More
Biomatik Corporation Phospho-IκB-β (Ser23) Colorimetric Cell-Based ELISA Kit
Supplier: Biomatik Corporation EKA50976
Phospho-IκB-β (Ser23) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IκB-β (Ser23) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15653. Gene ID: 4793. Target Synonyms: I-kappa-B-beta; IKB-B; IKBB; IkappaBbeta; NF-kappa-BIB; NF-kappaB inhibitor beta; NFKBIB; TR- interacting protein 9; TRIP9; Thyroid receptor interacting protein 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.