Learn More
Biomatik Corporation Phospho-Keratin 18 (Ser33) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50978
Encompass_Preferred
Phospho-Keratin 18 (Ser33) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Keratin 18 (Ser33) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05783. Gene ID: 3875. Target Synonyms: CK 18; CK18; CYK18; Cytokeratin endo B; K18; K1C18; KERD; KRT1-18; KRT18; Keratin; Keratin D; Keratin; type I cytoskeletal 18; cytokeratin 18; type I cytoskeletal 18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1194