Learn More
Biomatik Corporation Phospho-Shc (Tyr349) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50985
Encompass_Preferred
Phospho-Shc (Tyr349) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Shc (Tyr349) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29353. Gene ID: 6464. Target Synonyms: SH2 domain protein C1; SHC; SHC-transforming protein 1; SHCA; Src homology 2 domain-containing-transforming protein C1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1200